Skip to Content
Merck
All Photos(1)

Documents

S-075

Supelco

5-Fluoro PB-22 solution

100 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H21FN2O2
CAS Number:
Molecular Weight:
376.42
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule E (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IIA (Portugal)

concentration

100 μg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology
forensics and toxicology

format

single component solution

storage temp.

−20°C

InChI

1S/C23H21FN2O2/c24-13-4-1-5-15-26-16-19(18-10-2-3-11-20(18)26)23(27)28-21-12-6-8-17-9-7-14-25-22(17)21/h2-3,6-12,14,16H,1,4-5,13,15H2

InChI key

MBOCMBFDYVSGLJ-UHFFFAOYSA-N

General description

This certified Snap-N-Spike® Solution is suitable as starting material for calibrators or controls in synthetic cannabinoid GC/MS or LC/MS methods for urine drug testing, forensic analysis, or clinical toxicology applications. 5-Fluoro PB-22, also known as 5F-PB-22, is a synthetic cannabinoid first detected in Japanese herbal blends in 2013. The designer drug, a derivative of PB-22, is similar in structure to AM-2201.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F

Flash Point(C)

2.0 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service