Skip to Content
Merck
All Photos(1)

Key Documents

IMPN-007-04

Supelco

Naloxone N-oxide solution

100 μg/mL in acetonitrile: water (1:1), ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Naloxone N-oxide solution

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€214.00

€214.00


Available to ship on24 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 ML
€214.00

About This Item

Empirical Formula (Hill Notation):
C19H21NO5
CAS Number:
Molecular Weight:
343.37
UNSPSC Code:
41116107
NACRES:
NA.24

€214.00


Available to ship on24 April 2025Details


Request a Bulk Order

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in acetonitrile: water (1:1)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

Oc1ccc2C[C@@H]3[C@]4(O)CCC(=O)[C@@H]5Oc1c2[C@]45CC[N@@+]3([O-])CC=C

InChI

1S/C19H21NO5/c1-2-8-20(24)9-7-18-15-11-3-4-12(21)16(15)25-17(18)13(22)5-6-19(18,23)14(20)10-11/h2-4,14,17,21,23H,1,5-10H2/t14-,17+,18+,19-,20?/m1/s1

InChI key

GAUIBVINJLVYRY-QBFPIVRFSA-N

General description

Naloxone N-oxide is an impurity in pharmaceutical drug formulations of naloxone as well as a potential naloxone urinary metabolite. This certified solution standard is suitable for numerous LC or LC/MS applications from pharmaceutical testing and research to clinical toxicology or forensic analysis. Naloxone, sold under various trade names including Narcan®, Nalone, and Narcanti, is an opiate drug used to counter the effects of opiate overdose such as those involving heroin or morphine.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Narcan is a registered trademark of Endo Pharmaceuticals Inc.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service