Skip to Content
Merck
All Photos(1)

Key Documents

C-196

Supelco

Citalopram Impurity Mixture solution

15 μg/mL in methanol (each analyte), certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H21ClFNO · HBr
Molecular Weight:
414.74
UNSPSC Code:
41116107

grade

certified reference material

feature

Snap-N-Spike®/Snap-N-Shoot®

composition

Citalopram Hydrobromide, 15 μg/mL
Citalopram Related Compound A, 15 μg/mL
Citalopram Related Compound C, 15 μg/mL
Citalopram Related Compound D, 15 μg/mL
Citalopram Related Compound E, 15 μg/mL
Citalopram Related Compound F, 15 μg/mL
Citalopram Related Compound G, 15 μg/mL
Citalopram Related Compound H, 15 μg/mL

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

15 μg/mL in methanol (each analyte)

format

multi-component solution

storage temp.

−20°C

General description

Citalopram belongs to a class of drugs that are used as an antidepressant within the selective serotonin uptake inhibitor class. This certified solution standard contains multiple impurities, including Related Compounds A, C, D, E, F, G, and H as well as Citalopram itself. This standard can be used for multiple HPLC applications including pharmaceutical testing, research, and Monograph testing methods.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service