Skip to Content
Merck
All Photos(1)

Key Documents

870291P

Avanti

16:0 Folate Cap PE

Avanti Research - A Croda Brand 870291P, powder

Synonym(s):

1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine-N-(6-((folate)amino)hexanoyl) (sodium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€203.00

€203.00


In StockDetails


Request a Bulk Order

Select a Size

Change View
1 MG
€203.00

About This Item

Empirical Formula (Hill Notation):
C62H100N9Na2O14P
CAS Number:
Molecular Weight:
1272.46
UNSPSC Code:
12352211
NACRES:
NA.25

€203.00


In StockDetails


Request a Bulk Order

form

powder

packaging

pkg of 1 × 1 mg (870291P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870291P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCN([H])C(CCCCCNC(CC[C@H](NC(C1=CC=C(NCC(C=N2)=NC3=C2N=C(N)NC3=O)C=C1)=O)C([O-])=O)=O)=O)=O)(OC(CCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O.[Na+].[Na+]

General description

16:0 Folate Cap PE, also known as 1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine-N-(6-((folate)amino)hexanoyl) (PE-FA), is a head group modified lipid.

Application

16:0 Folate Cap PE or 1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine-N-(6-((folate)amino)hexanoyl) has been used to synthesize functional polymer dots.[1][2]

Packaging

5 mL Amber Glass Screw Cap Vial (870291P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High resolution tracking of macrophage cells in deep organs and lymphatics using fluorescent polymer dots
Tang S, et al.
Royal Society of Chemistry Advances, 9(19), 10966-10975 (2019)
Yufan Zhang et al.
Molecular imaging and biology, 21(6), 1026-1033 (2019-03-17)
Probe-based confocal laser endomicroscopy (pCLE) is a novel technique allowing real-time and high-resolution imaging in vivo. It provides microscopic images and increases the penetration depth of tissues compared with conventional white light endoscopy. The aim of the present study was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service