Skip to Content
Merck
All Photos(1)

Key Documents

860468P

Avanti

N-C24:1-desoxymethylsphingosine

Avanti Research - A Croda Brand 860468P, powder

Synonym(s):

N-nervonoyl-1-desoxymethylsphingosine (m17:1/24:1)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€333.00

€333.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€333.00

About This Item

Empirical Formula (Hill Notation):
C41H79NO2
CAS Number:
Molecular Weight:
618.07
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€333.00


Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 1 mg (860468P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860468P

lipid type

bioactive lipids
sphingolipids

shipped in

dry ice

storage temp.

−20°C

General description

Ceramides are generated by the acylation of the sphingoid bases, sphingosine.[1]

Biochem/physiol Actions

Deoxydihydro ceramide (deoxyDHCer), nervonoyl (C24:1) is present in the mammals along with palmitoyl (C16) dihydroceramide. In general, deoxyDHCer modulates the cellular membrane properties and may contribute to its biological functionality.[2]

Packaging

5 mL Amber Glass Screw Cap Vial (860468P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sphingolipid biosynthesis in man and microbes
Harrison PJ, et al.
Natural Product Reports, 35(9), 921-954 (2018)
Biophysical properties of novel 1-deoxy-(dihydro) ceramides occurring in mammalian cells
Jimenez-Rojo N, et al.
Biophysical Journal, 107(12), 2850-2859 (2014)
Junliang Wan et al.
Journal of agricultural and food chemistry, 67(46), 12953-12961 (2019-10-23)
Most common sphingolipids are comprised of "typical" sphingoid bases (sphinganine, sphingosine, and structurally related compounds) and are produced via the condensation of l-serine with a fatty acyl-CoA by serine palmitoyltransferase. Some organisms, including mammals, also produce "atypical" sphingoid bases that
Sarah T Pruett et al.
Journal of lipid research, 49(8), 1621-1639 (2008-05-24)
"Sphingosin" was first described by J. L. W. Thudichum in 1884 and structurally characterized as 2S,3R,4E-2-aminooctadec-4-ene-1,3-diol in 1947 by Herb Carter, who also proposed the designation of "lipides derived from sphingosine as sphingolipides." This category of amino alcohols is now

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service