Skip to Content
Merck
All Photos(2)

Key Documents

850851C

Avanti

16:0 Monomethyl PE

1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine-N-methyl, chloroform

Synonym(s):

1,2-dihexadecanoyl-sn-glycero-3-phosphoethanolamine-N-methyl ; 110743

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H76NO8P
CAS Number:
Molecular Weight:
705.99
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (850851C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850851C

concentration

10 mg/mL (850851C-25mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC[N+](C)([H])[H])=O)(OC(CCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O

InChI

1S/C38H76NO8P/c1-4-6-8-10-12-14-16-18-20-22-24-26-28-30-37(40)44-34-36(35-46-48(42,43)45-33-32-39-3)47-38(41)31-29-27-25-23-21-19-17-15-13-11-9-7-5-2/h36,39H,4-35H2,1-3H3,(H,42,43)

InChI key

QSBINWBNXWAVAK-UHFFFAOYSA-N

General description

16:0 Monomethyl phosphoethanolamine (PE) is an alkyl phosphate derivative.

Application

16:0 Monomethyl PE has been used as an internal calibration standard in liquid chromatography tandem mass spectrometry (LC-MS/MS) for the extraction of phospholipids in foods and tissues.[1]

Packaging

5 mL Clear Glass Sealed Ampule (850851C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Validation of an LC-MS/MS method for the quantification of choline-related compounds and phospholipids in foods and tissues
Xiong Y, et al.
Journal of Chromatography. B, Analytical Technologies in the Biomedical and Life Sciences, 911, 170-179 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service