Skip to Content
Merck
All Photos(1)

Key Documents

850561P

Avanti

Popular Detergent Kit

Avanti Research - A Croda Brand

Synonym(s):

Detergent Evaluation Kit

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€650.00

€650.00


Available to ship on14 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 EA
€650.00

About This Item

UNSPSC Code:
12161902
NACRES:
NA.28

€650.00


Available to ship on14 April 2025Details


Request a Bulk Order

Assay

99% (TLC)

form

powder

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

General description

The Popular Detergent Kit is designed to allow for the convenient evaluation of 6 unique detergent reagents for their ability to influence membrane protein solubilization, purification, and stabilization during crystal growth. This kit provides a great starting point for determining which detergent or which class of detergent will work best for a particular membrane protein.

Packaging

20ML Clear Glass Screw Cap Vial for each component (850561P-1EA)
Package contains 1G of each: n-octyl-β-D-glucopyranoside, MAPCHO-12, GDN, MEGA-9, n-dodecyl- β-D-maltoside (DDM), CHAPS

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service