Skip to Content
Merck
All Photos(2)

Key Documents

810128C

Avanti

12:0 Lyso NBD PC

Avanti Research - A Croda Brand 810128C

Synonym(s):

1-{12-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]dodecanoyl}-2-hydroxy-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H44N5O10P
CAS Number:
Molecular Weight:
617.63
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (LPC; may contain up to 10% of the 2-LPC isomer, TLC)

form

liquid

packaging

pkg of 1 × 1 mL (810128C-1mg)
pkg of 5 × 1 mL (810128C-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810128C

concentration

1 mg/mL (810128C-1mg)
1 mg/mL (810128C-5mg)

lipid type

fluorescent lipids

shipped in

dry ice

storage temp.

−20°C

Application

12:0 Lyso NBD PC may be used as a substrate to measure the enzymatic activity of recombinant human lysophosphatidylcholine acyltransferase 3 (rhLPCAT3)[1] and to study the function of lysophosphatidic acid (LPA) as an inhibitor in the autotaxin (ATX) catalysis.[2]

Packaging

5 mL Clear Glass Sealed Ampule (810128C-1mg)
5 mL Clear Glass Sealed Ampule (810128C-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fernando Salgado-Polo et al.
The Journal of biological chemistry, 293(37), 14312-14327 (2018-07-22)
Autotaxin (ATX) is a secreted glycoprotein and the only member of the ectonucleotide pyrophosphatase/phosphodiesterase family that converts lysophosphatidylcholine (LPC) into lysophosphatidic acid (LPA). LPA controls key responses, such as cell migration, proliferation, and survival, implicating ATX-LPA signaling in various (patho)physiological
Lauren P Saunders et al.
The Journal of biological chemistry, 286(34), 30130-30141 (2011-07-02)
Autotaxin (ATX) is a secreted lysophospholipase D that hydrolyzes lysophosphatidylcholine (LPC) into lysophosphatidic acid (LPA), initiating signaling cascades leading to cancer metastasis, wound healing, and angiogenesis. Knowledge of the pathway and kinetics of LPA synthesis by ATX is critical for
Xinming Du et al.
FEBS open bio, 9(10), 1734-1743 (2019-08-04)
Lysophosphatidylcholine acyltransferase 3 (LPCAT3) is an important enzyme in phospholipid remodeling, a process that influences the biophysical properties of cell membranes and thus cell function. Multiple lines of evidence suggest that LPCAT3 is involved in several diseases, including atherosclerosis, non-alcoholic
Stephen M Baumler et al.
Langmuir : the ACS journal of surfaces and colloids, 33(12), 2986-2992 (2017-03-08)
The translational diffusion dynamics of the zwitterionic lipid 1,2-dimyristoyl-sn-glycero-3-phosphocholine (DMPC) at a planar phosphorylated support surface containing metal ions (Mg2+, Ca2+, Ba2+, Ni2+, Zn2+, Cd2+, Zr4+) was investigated using X-ray photoelectron spectroscopy (XPS) and fluorescence recovery after photobleaching (FRAP). Fluorescence

Articles

The critical micelle concentration (CMC) can be approximately defined as the lipid monomer concentration at which appreciable amounts (>5% of total) of micellar aggregates first begin to appear in the equilibrium: nM1<=>Mn

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service