700261P
Avanti
Glycodehydrocholic acid
Avanti Research™ - A Croda Brand
Synonym(s):
5ß-cholanic acid-3,7,12-trione N-(2-carboxymethyl)-amide
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
form
solid
packaging
package of 1 × 10 mg
manufacturer/tradename
Avanti Research™ - A Croda Brand
lipid type
bile acids
shipped in
dry ice
storage temp.
−20°C
Packaging
5 mL Amber Glass Screw Cap Vial (700261P-10MG)
Legal Information
Avanti Research is a trademark of Avanti Polar Lipids, LLC
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service