Skip to Content
Merck
All Photos(1)

Key Documents

700137P

Avanti

7β,27-dihydroxycholesterol-d6

Avanti Research - A Croda Brand 700137P, powder

Synonym(s):

7β,27-dihydroxycholesterol-d6

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€884.00

€884.00


Available to ship on01 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 MG
€884.00

About This Item

Empirical Formula (Hill Notation):
C27H40D6O3
CAS Number:
Molecular Weight:
424.69
MDL number:
UNSPSC Code:
41141804
NACRES:
NA.25

€884.00


Available to ship on01 April 2025Details


Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (700137P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 700137P

shipped in

dry ice

storage temp.

−20°C

General description

7β,27-dihydroxycholesterol-d6 is a deuterated derivative of 7β,27-dihydroxycholesterol. 7β,27-dihydroxycholesterol is a substrate for enzyme 11β-hydroxysteroid dehydrogenases (11β-HSDs).[1]

Application

7β,27-dihydroxycholesterol-d6 has been used as an internal standard in ultra-performance liquid chromatography - tandem mass spectrometer (UPLC-MS/MS).[1]

Biochem/physiol Actions

7β,27-dihydroxycholesterol (7β27OHC) serves as a ligand for retinoid-related orphan receptor γ (RORγ). 7β27OHC is also involved in smoothened (Smo) functionality.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (700137P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katharina R Beck et al.
Journal of lipid research, 60(9), 1535-1546 (2019-07-06)
Oxysterols previously were considered intermediates of bile acid and steroid hormone biosynthetic pathways. However, recent research has emphasized the roles of oxysterols in essential physiologic processes and in various diseases. Despite these discoveries, the metabolic pathways leading to the different

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service