Skip to Content
Merck
All Photos(1)

Key Documents

640020P

Avanti

DMABA Kit

Avanti Research - A Croda Brand

Synonym(s):

DMABA Kit

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€379.00

€379.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
€379.00

About This Item

UNSPSC Code:
12161503
NACRES:
NA.28

€379.00


Check Cart for Availability

Request a Bulk Order

Assay

>99%

form

powder

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

General description

The DMABA NHS ester reagents were developed by Dr. Robert Murphy′s laboratory at the University of Colorado in order to create PE derivatives where all subclasses and potentially oxidized products could be universally detected using a common precursor ion in the positive ion mode by Mass Spectrometry.

Biochem/physiol Actions

Deuterium enriched 4-(dimethylamino)benzoic acid (DMABA) N-hydroxysuccinimide (NHS) ester reagent reacts with the with the primary amine group of PE lipids.[1]

Packaging

Kit contains: 1mg each of (D0, D4, D6 and D10)-4(dimethylamino)-benzoic acid N-hydroxysuccinimide ester

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Kit Components Also Available Separately

Product No.
Description
SDS

  • 640000PDMABA-d0 NHS Ester, Avanti Research - A Croda BrandSDS

  • 640004PDMABA-d4 NHS Ester, Avanti Research - A Croda BrandSDS

  • 640006PDMABA-d6 NHS Ester, Avanti Research - A Croda BrandSDS

  • 640010PDMABA-d10 NHS Ester, Avanti Research - A Croda BrandSDS

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karin A Zemski Berry et al.
Analytical chemistry, 81(16), 6633-6640 (2010-03-27)
A set of four (D(0), D(4), D(6), and D(10)) deuterium enriched 4-(dimethylamino)benzoic acid (DMABA) N-hydroxysuccinimide (NHS) ester reagents was developed that react with the primary amine group of glycerophosphoethanolamine (PE) lipids to create derivatives where all subclasses of DMABA labeled

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service