Skip to Content
Merck
All Photos(2)

Key Documents

330274W

Avanti

Taurochenodeoxycholic acid-D4 sodium salt

Avanti Research - A Croda Brand

Synonym(s):

5ß-cholanic acid-3α,7α-diol N-(2-sulphoethyl)-amide-d4, sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€223.00

€223.00


Available to ship on16 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€223.00

About This Item

Empirical Formula (Hill Notation):
C26H40D4NNaO6S
CAS Number:
Molecular Weight:
525.71
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.12

€223.00


Available to ship on16 April 2025Details


Request a Bulk Order

Assay

>99% (MS)

form

methanol solution

packaging

package of 1 × 100 μg

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

100 μg/mL in methanol

shipped in

dry ice

storage temp.

−20°C

Packaging

2 mL Amber Glass Sealed Ampule (330274W-100UG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service