Skip to Content
Merck
All Photos(1)

Key Documents

ATE443545536

AstaTech, Inc.

5-FLUOROPYRAZINE-2-CARBOXYLIC ACID

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
142.09
MDL number:

Iupac Name

5-fluoro-2-pyrazinecarboxylic acid

Inchi Code

1S/C5H3FN2O2/c6-4-2-7-3(1-8-4)5(9)10/h1-2H,(H,9,10)

InChI key

PVJHQFMTUXTQNZ-UHFFFAOYSA-N

storage temp.

2-8 C

Purity

95%

Shipping Temperature

Normal

Physical Form

White to Yellow Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=56807

Signal Word

Warning

Hazard Statements

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=70428


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library