Skip to Content
Merck
All Photos(1)

Key Documents

ATE398811585

AstaTech, Inc.

6-BROMOBENZO[B]THIOPHENE-3-CARBALDEHYDE

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
241.11
MDL number:

Iupac Name

6-bromobenzo[b]thiophene-3-carbaldehyde

Inchi Code

1S/C9H5BrOS/c10-7-1-2-8-6(4-11)5-12-9(8)3-7/h1-5H

InChI key

PBCQAUIPFVKGOW-UHFFFAOYSA-N

storage temp.

2-8 C

Purity

95%

Shipping Temperature

Normal

Physical Form

Pale-yellow to Yellow-brown Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=18838

Signal Word

Warning

Hazard Statements

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=82318


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library