APOH04239E0F
Apollo Scientific Ltd
4-Iodo-2-methoxybenzaldehyde
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
4-iodo-2-methoxybenzaldehyde
Inchi Code
1S/C8H7IO2/c1-11-8-4-7(9)3-2-6(8)5-10/h2-5H,1H3
InChI key
ZDNOHFJWXJKIJF-UHFFFAOYSA-N
storage temp.
Ambient Temperature
Country of Origin
GB
Shipping Temperature
Ambient Temperature
Physical Form
Solid
Product Link
https://store.apolloscientific.co.uk/product/4-iodo-2-methoxybenzaldehyde
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.