Key Documents
Z755958
Brady® BMP®21-LAB hand-held label printer
AC/DC input 220 V AC, EuroPlug
Synonym(s):
label machine, label printers, labels printer, printer labels
About This Item
UNSPSC Code:
55121611
NACRES:
NB.35
packaging
pkg of 1 ea
manufacturer/tradename
Brady BMP21-LAB-KIT EU
General description
The BMP®21 Hand-held Label Printer, eliminate guesswork and enjoy fast and easy formatting of commonly used label types for marking wire and cable, terminal blocks, patch panels and much more. You can be confident your Brady labels will remain stuck for years. Create clear, legible labels with industrial-strength staying power to withstand even the harshest environments. They won′t fall off, smear or fade.
Simple to operate and quick to print, it′s a labelling tool specifically designed for industrial environments like voice/datacom, electrical and general industrial. Print one label, knowing it′ll adhere and stay legible for the years to come.
The BMP21 is a labelling solution that leads to increased productivity, supreme accuracy and unrivalled quality for a safer and a more efficient workplace.
Features and Benefits:
  • Hard wearing design for industrial environments
  • 203 dpi thermal transfer print
  • Large LCD Display (3 lines of text, 1 line of icons)
  • Magnet and pole strap for one-handed operation
  • Flashlight accessory for use in confined and low light areas
  • Prints on a varity of materials including Continuous Labels
  • Automatic data serialisation and built-in graphics and barcodes
  • Access to 80 different symbols, including 48 general identification symbols
  • Ergonomic dual cutter buttons that hold the label until you need it
  • Six languages (English, Spanish, German, French, Portuguese, Dutch)
  • Barcodes (3 of 9 and 128)
  • Integrated Smart Cell technology
  • Font Autosizing
Legal Information
BMP is a registered trademark of Brady Worldwide, Inc.
Brady is a registered trademark of Brady Worldwide, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)