Key Documents
Z743202
Julabo® bath carrier trays for SW22/SW23
spring clamps for use with universal basic tray
Select a Size
1 EA
€77.00
€77.00
Check Cart for Availability
Select a Size
Change View
1 EA
€77.00
About This Item
UNSPSC Code:
41103706
NACRES:
NB.77
€77.00
Check Cart for Availability
manufacturer/tradename
Julabo 8970631
General description
Spring clamps set for use with basic stainless steel tray Z743203 (8970630). Set includes 12 spring clamps 135 mm long and 5 spring clamps 190 mm long. For use with Julabo® shaking water bath models SW22, SW23.
Legal Information
Julabo is a registered trademark of Julabo USA, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)