Documents
Z740177
Millipore
Amicon® Ultra-0.5
Ultracel-50 membrane, 50 kDa
About This Item
UNSPSC Code:
41104816
material
regenerated cellulose membrane
styrene-butadiene housing
sterility
non-sterile
feature
50 kDa
packaging
pack of 24 ea
manufacturer/tradename
Millipore UFC505024
tube size
~1.5 mL
working volume
0.5 mL
pore size
50 kDa MWCO
Legal Information
Amicon is a registered trademark of Merck KGaA, Darmstadt, Germany
Visit the Document Library
Esteban Cruz et al.
Cancers, 11(6) (2019-06-27)
Nanoparticle carriers offer the possibility of enhanced delivery of therapeutic payloads in tumor tissues due to tumor-selective accumulation through the enhanced permeability and retention effect (EPR). Gold nanoparticles (AuNP), in particular, possess highly appealing features for development as nanomedicines, such