Key Documents
Z691038
Quickfit® flat flange lids for reaction flasks
flange bore 75 mm, center joint: ST/NS 34/35
Synonym(s):
DA5/75
Select a Size
Z691038-1EA
€151.00
€151.00
Estimated to ship on29 May 2025
Select a Size
Change View
Z691038-1EA
€151.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
€151.00
Estimated to ship on29 May 2025
material
borosilicate glass
Joint
center joint: ST/NS34/35
sterility
non-sterile
packaging
case of 1
manufacturer/tradename
Quickfit DA5/75
availability
not available in USA and Canada
flange bore
75 mm
General description
Single and multi-socket lids for reaction flasks for simple extraction, reflux or for the introduction of a variety of stirrers, dropping funnels, temperature probes etc.
Use clip to secure lid to flasks, purchase separately.
Legal Information
Quickfit is a registered trademark of SciLabware Limited
Choose from one of the most recent versions:
Certificates of Analysis (COA)