Key Documents
Z687944
Levochem ethylene-vinyl-alcohol gloves
size 10
About This Item
UNSPSC Code:
46181504
size
10
feature
standard type EN 374 (cat. III)
standard type EN 388 (cat. III)
manufacturer/tradename
KCL 761-10
packaging
pkg of Packaged in boxes of 10 pairs and cases of 10 boxes
L × thickness
360-400 mm × 0.07 mm , +/- 0.01 mm
General description
Levochem is made of ethylene-vinyl-alcohol (EVOH) and polyethylene welded 3-layer sandwich system. It offers high resistance against a variety of hazardous substance groups and can be worn on its own or under another glove.
  • Very good protection against variety of hazardous substance groups
  • Can be worn under gloves
  • Excellent for use in emergency spill applications
  • Ambidextrous
  • CE compliant
Choose from one of the most recent versions:
Certificates of Analysis (COA)