Key Documents
Z557765
Sigma-Aldrich
Aldrich® swivel McLeod vacuum gauge
replacement parts kit
About This Item
UNSPSC Code:
41112406
NACRES:
NB.79
General description
Contains a set replacement o-ring seals and plastic nuts for the swiveling connection that joins the two glass components.
Made with permanent linear and square root ceramic scales on a white background, high-vacuum O-ring seals, precision bore capillaries, Delrin® plastic and black anodized aluminum parts. Accurate to ±3% of reading or 1mm of scale division, whichever is greater. Gauges include operating instructions but do not include oil or mercury fill. Swivel arrangement permits rigid connection to the vacuum system with the scales vertical at all times.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Delrin is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)