Key Documents
Z531170
Sigma-Aldrich
Aldrich® coiled condenser
unjacketed, L 400 mm, joint: ST/NS 29/32
About This Item
UNSPSC Code:
41121800
Joint
joint: ST/NS29/32
L
400 mm
General description
The coiled condenser circulates coolant through the coil. The jacketed version provides additional cooling capacity by allowing vapors to condense on the inner wall of the jacket.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)