Z503851
Laboratory brushes with PP handles
Funnel/beaker, brush diam. × L 2 1/4 in. × 6 in.
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41100000
NACRES:
NB.35
Recommended Products
manufacturer/tradename
Justrite 5411
Overall L
16 in.
brush diam. × L
2 1/4 in. × 6 in.
Looking for similar products? Visit Product Comparison Guide
General description
Durable black nylon filament brushes with tufted ends have white, textured-grip, molded PP handles. Brushes will not scratch or mar equipment, absorb moisture, chemicals, or oils.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service