Skip to Content
Merck
All Photos(2)

Key Documents

Z404144

IKA® VRX basic Vibrax® orbital shaker attachments

Erlenmeyer flask attachment, for Erlenmeyer flasks (2 flasks up to 500mL), 1/cs

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103811
NACRES:
NB.73

packaging

pack of 1

manufacturer/tradename

IKA 0000910400

H × W × D

60 mm × 265 mm × 136 mm

Looking for similar products? Visit Product Comparison Guide

General description

Note: Securely holds Erlenmeyer flasks (2 flasks up to 500mL).

Legal Information

IKA is a registered trademark of IKA-Werke GmbH & Co. KG
Vibrax is a registered trademark of IKA-Werke GmbH & Co. KG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service