Key Documents
Z401870
MSA Defender + faceshield
clear polycarbonate visor
About This Item
UNSPSC Code:
46181802
Pricing and availability is not currently available.
material
clear polycarbonate visor
manufacturer/tradename
MSA 488132
availability
not available in EU
size
8 in. × 16 in. × 0.04 in.
General description
  • Lightweight, flexible headbands adjust to accommodate head sizes 61/2 to 8
  • Effective against most flying fragments, particles, splashes, and similar hazards
  • Optional nitrometer chin guard protects against upsweep of particles and splash
  • Use with approved spectacles or goggles
  • Assemblies comply with ANSI Z87.1
Order Defender + headgear assembly, visor and chin guard separately.
Choose from one of the most recent versions:
Certificates of Analysis (COA)