Key Documents
Z341894
IKA® dish attachment for KS 130 basic and control shakers
AS 130.3 dish attachment
Select a Size
1 EA
€125.00
€125.00
Estimated to ship on08 May 2025
Select a Size
Change View
1 EA
€125.00
About This Item
UNSPSC Code:
41121500
NACRES:
NB.73
€125.00
Estimated to ship on08 May 2025
material
foil (slip-resistant)
packaging
pkg of 1 ea
manufacturer/tradename
IKA 3120000
General description
Note: For smooth shaking operations in the low viscosity range, e.g. Petri dishes or culture bottles. With integrated slip-resistant foil (PP).
Legal Information
IKA is a registered trademark of IKA-Werke GmbH & Co. KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)