Z340510
Binder drying oven FD series
FD 056, AC/DC input 230 V AC
Synonym(s):
benchtop drying oven, drying oven
Select a Size
€1,980.00
Estimated to ship on12 May 2025
Select a Size
About This Item
€1,980.00
Estimated to ship on12 May 2025
Recommended Products
AC/DC input
230 V
feature
CE compliant
timer Electronic 0-99.59hr
packaging
pack of 1 ea
manufacturer/tradename
FD 056
availability
not available in USA and Canada
parameter
10 C above room temperature-30 °C temp. range
inner chamber volume
60 L
inside L × W × H
565 mm × 560 mm × 625 mm
weight
40 kg
Looking for similar products? Visit Product Comparison Guide
General description
- Hot air sterilizers with forced convection
- 2 chrome plated shelves
- Temperature safety device CI.2
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service