Key Documents
Z284181
Spinvane® magnetic stirring bar
for centrifuge tubes
Synonym(s):
Stir Bar, Stirbar, Stirring Bar
About This Item
UNSPSC Code:
41103806
material
PTFE-coated (Half round tapered)
PTFE-coated
packaging
pack of 5 ea
manufacturer/tradename
Bel-Art F37137-0000
bar W × H × diam.
9.7/6.1 mm × 23.0 mm × 9.9 mm
General description
  • Spinvane® magnetic stirring bars are ideal for small conical-shaped tubes and micro vials.
  • The triangle-shaped polytetrafluoroethylene (PTFE) coated vane features a cross-mounted molded-in Alnico magnet for effective stirring.
  • Autoclavable.
Bars designed for narrow stirring applications.
Legal Information
Spinvane is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)