Key Documents
Z179973
Supelco
Filter column for Michel-Miller columns
About This Item
UNSPSC Code:
12000000
NACRES:
SB.52
manufacturer/tradename
Ace Glass 581323
technique(s)
LPLC: suitable
L × I.D.
85 mm × 8 mm
General description
Assures only clean solvent enters M-M column. Position in-line between pump and column using Ace-Thred #7 end-fitting adapters and 1/4-28 tubing adapters. Connections are Ace-Thred #7. Order adapters and filter material separately.
Choose from one of the most recent versions:
Certificates of Analysis (COA)