Key Documents
Z172952
Sigma-Aldrich
Aldrich® pour-out adapter
male joint: ST/NS 14/20
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
Joint
male joint: ST/NS14/20
General description
For pouring from flasks or funnels without pouring over ground joint.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)