Key Documents
WHA230010063
Whatman® Benchkote® surface protector for ÄKTA® Start
pack of 10 ea
About This Item
UNSPSC Code:
41122300
NACRES:
NB.24
Pricing and availability is not currently available.
material
(Absorbent paper and laminated polyethylene )
packaging
pack of 10 ea
manufacturer/tradename
Cytiva
Whatman 2300-10063
Related Categories
General description
Why Benchkote sheet for lab ÄKTA systems?
  • Cover the top buffer tray with Benchkote sheet and protect from buffer spillages and salt deposits.
  • Simplify daily and weekly preventive maintenance of the top buffer tray of your ÄKTA systems Benchkote sheet
  • Smooth, absorbent Whatman paper quickly absorbs spills.
  • Laminated polyethylene layer prevents flow-through to the working surface.
  • Fits the top buffer tray on ÄKTA avant, ÄKTA pure and ÄKTA start protein purification systems.
  • Offered in three convenient pack sizes: 10/pk, 25/pk and 50/pk sheets.
  • Convenient and easy to use.
Benchkote sheets are manufactured of absorbent,
impermeable material designed to protect lab surfaces against hazardous spills. The high-quality, smooth, and absorbent Whatman® paper, quickly absorbs liquid spills, and laminated polyethylene layer prevents flow-through to the working surface.
Features and Benefits
  • Cover the top buffer tray with Benchkote sheet and protect from buffer spillages and salt deposits.
  • Simplify daily and weekly preventive maintenance of the top buffer tray of your ÄKTA systems
  • Smooth, absorbent Whatman paper quickly absorbs spills.
  • Laminated polyethylene layer prevents flow-through to the working surface.
  • Fits the top buffer tray on ÄKTA go, ÄKTA avant, ÄKTA pure and ÄKTA start protein purification systems.
  • Offered in three convenient pack sizes: 10/pk, 25/pk and 50/pk sheets.
  • Convenient and easy to use.
Legal Information
Benchkote is a registered trademark of Cytiva
Whatman is a registered trademark of Cytiva
ÄKTA is a registered trademark of Cytiva
Choose from one of the most recent versions:
Certificates of Analysis (COA)