WHA2017009
Whatman® Antibiotic Assay Discs
diam. 9 mm
Synonym(s):
Z741329
All Photos(1)
About This Item
Recommended Products
packaging
pkg of 1000 ea
manufacturer/tradename
Whatman 2017-009
Whatman Article No. 28418661 (US reference)
diam.
9 mm
Related Categories
General description
For determining the type of causal agent of infectious diseases and for checking their sensitivity to antibiotics and chemotherapeutic agents in vitro by means of the inhibition zone determination method.
Features and Benefits
- The antibiogram allows rational and selective chemotherapy.
- Test discs can be coated with chemotherapeutic agents, placed on the innoculated nutrient agar and incubated.
- The size of the inhibition zone is a measure for the effectiveness of the substances.
Other Notes
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Legal Information
Whatman is a registered trademark of Cytiva