Documents
WHA10498761
Whatman® multiple vacuum filtration device
3-place, 3-place manifold, stainless steel
Synonym(s):
Z752177
About This Item
UNSPSC Code:
41104900
NACRES:
NB.24
material
stainless steel
packaging
pk of 1
manufacturer/tradename
Whatman 10498761
Whatman Article No. 28416533 (US reference)
General description
Stainless steel filter funnel manifold, for microbiology monitors and analytical funnels.
Features and Benefits
  • The stainless steel manifold for three or six filtration units is fitted with stainless steel units.
  • The apparatus can be autoclaved and sterilized by dry heat at up to 180°C.
  • Suitable only for vacuum operation.
  • If flushing tubes are used, do not exceed 1.3 bar (300 mbar over-pressure).
Other Notes
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Legal Information
Whatman is a registered trademark of Cytiva