Skip to Content
Merck
All Photos(1)

Key Documents

W504904

Sigma-Aldrich

Butyrophenone

≥99%

Synonym(s):

1-Benzoylpropane, 1-Phenyl-1-butanone, NSC 8463, Phenyl propyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COCH2CH2CH3
CAS Number:
Molecular Weight:
148.20
Beilstein:
508305
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
7.193
NACRES:
NA.21

biological source

synthetic

Assay

≥99%

refractive index

n20/D 1.520 (lit.)

bp

228-230 °C (lit.)

mp

11-13 °C (lit.)

density

1.021 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

camphoraceous

SMILES string

CCCC(=O)c1ccccc1

InChI

1S/C10H12O/c1-2-6-10(11)9-7-4-3-5-8-9/h3-5,7-8H,2,6H2,1H3

InChI key

FFSAXUULYPJSKH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

190.4 °F

Flash Point(C)

88 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fabrice Gritti et al.
Journal of chromatography. A, 1410, 118-128 (2015-08-10)
An original method is proposed for the accurate and reproducible measurement of the time-based dispersion properties of short L< 50cm and narrow rc< 50μm tubes at mobile phase flow rates typically used in very high-pressure liquid chromatography (vHPLC). Such tubes
Fabrice Gritti et al.
Journal of chromatography. A, 1420, 54-65 (2015-10-17)
The impact of the column hardware volume (≃ 1.7 μL) on the optimum reduced plate heights of a series of short 2.1 mm × 50 mm columns (hold-up volume ≃ 80-90 μL) packed with 1.8 μm HSS-T3, 1.7 μm BEH-C18
Ruben De Pauw et al.
Journal of chromatography. A, 1420, 129-134 (2015-10-20)
A new system is proposed for applying fast temperature gradients in liquid chromatography. It consists of a 0.7 mm × 150 mm fused-silica column coated with a 50 μm Nickel-layer, which is connecting with a power source and a temperature
Zsolt Székelyhidi et al.
Bioorganic & medicinal chemistry letters, 15(13), 3241-3246 (2005-06-01)
SR protein-specific kinase-1 (SRPK-1) has been identified as a validated target for hepatitis B virus (HBV). A series of novel tricyclic quinoxaline derivatives was designed and synthesised as potential kinase inhibitory antiviral agents and was found to be active and
Nikola Vaňková et al.
Journal of chromatography. A, 1409, 152-158 (2015-07-29)
The effect of gradient steepness on the kinetic performance limits and peak compression effects has been assessed in gradient mode for the separation of phenol derivatives using columns packed with 2.6μm core-shell particles. The effect of mobile-phase velocity on peak

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service