Skip to Content
Merck
All Photos(1)

Key Documents

W423601

Sigma-Aldrich

Ethylamine solution

70 wt. % in H2O

Synonym(s):

Aminoethane, Monoethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5NH2
CAS Number:
Molecular Weight:
45.08
FEMA Number:
4236
Beilstein:
505933
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
11.015

biological source

synthetic

concentration

70 wt. % in H2O

refractive index

n20/D 1.384

bp

17 °C/760 mmHg

mp

-74 °C

density

0.81 g/mL at 20 °C
0.810 g/mL at 25 °C

application(s)

flavors and fragrances

SMILES string

CCN

InChI

1S/C2H7N/c1-2-3/h2-3H2,1H3

InChI key

QUSNBJAOOMFDIB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Flam. Liq. 2 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

71.6 °F

Flash Point(C)

22 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuying Shuai et al.
Journal of the science of food and agriculture, 90(15), 2563-2567 (2010-08-26)
Theanine, a unique amino acid found almost exclusively in tea plants, has various favourable physiological and pharmacological functions in humans. Gamma-glutamyltranspeptidase (GGT, EC 2.3.2.2) is considered to be the most effective enzyme for the production of theanine. In fact, GGT
Câline Christine et al.
Organic & biomolecular chemistry, 10(46), 9183-9190 (2012-10-23)
The synthesis of a phosphonated acyclic bifunctional chelate L* for the labeling of biomaterial is described. L* is based on a pyridine backbone, functionalized in ortho positions by aminomethyl-bis-methylphosphonic acids, and, in the para position, by a side chain containing
Song Yang et al.
Journal of chromatography. A, 1216(15), 3280-3289 (2009-03-10)
Complementary methods using liquid chromatography-tandem quadrupole mass spectrometry (LC-MS/MS) and comprehensive two-dimensional gas chromatography-time-of-flight mass spectrometry (GCxGC-TOF-MS) were developed and applied to determine targeted metabolites involved in central carbon metabolism [including tricarboxylic acid cycle, serine cycle, ethylmalonyl-coenzyme A (ethylmalonyl-CoA) pathway
Lisa Vasicek et al.
Analytical chemistry, 81(19), 7876-7884 (2009-09-03)
Electron transfer dissociation (ETD) has proven to be a promising new ion activation method for proteomics applications due to its ability to generate c- and z-type fragment ions in comparison to the y- and b-type ions produced upon the more
Tracy A LeGreve et al.
The journal of physical chemistry. A, 113(2), 399-410 (2008-12-23)
Neutral serotonin-(H(2)O)(n) clusters with n = 1,2 have been studied under jet-cooled conditions using a combination of resonant two-photon ionization (R2PI), UV-UV hole-burning (UVHB), and resonant ion-dip infrared (RIDIR) spectroscopy. Serotonin (5-hydroxytryptamine, SERO) is a close analogue of tryptamine, differing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service