Skip to Content
Merck
All Photos(1)

Key Documents

W382604

Sigma-Aldrich

2,5-Dihydroxy-1,4-dithiane

mixture of isomers, ≥97%, FG

Synonym(s):

1,4-Dithiane-2,5-diol, 2,5-Dihydroxy-1,4-dithiane, Mercaptoacetaldehyde dimer

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O2S2
CAS Number:
Molecular Weight:
152.24
FEMA Number:
3826
Beilstein:
104402
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
15.134
grade:
FG
Halal
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

grade

FG
Halal

reg. compliance

EU Regulation 1334/2008 & 872/2012

Assay

≥97%

mp

130 °C (dec.) (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

bread; meaty; sulfurous; tomato

storage temp.

2-8°C

SMILES string

OC1CSC(O)CS1

InChI

1S/C4H8O2S2/c5-3-1-7-4(6)2-8-3/h3-6H,1-2H2

InChI key

YUIOPHXTILULQC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,5-Dihydroxy-1,4-dithiane is a sulfur-containing compound that is reported to show phytogrowth-inhibitory activities on the root growth of some Brassica species.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1A - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Inamori et al.
Chemical & pharmaceutical bulletin, 38(1), 243-245 (1990-01-01)
2,5-Dihydroxy-1,4-dithiane (I) and its derivatives (II-IV) showed rather marked inhibitory activities on the growth of the roots of two plant species. All compounds tested had phytogrowth-inhibitory activities. These compounds markedly inhibited the growth of the two plant species at the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service