Skip to Content
Merck
All Photos(1)

Key Documents

W368318

Sigma-Aldrich

Ethyl 3-oxohexanoate

≥98%

Synonym(s):

Ethyl butyrylacetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2COCH2COOC2H5
CAS Number:
Molecular Weight:
158.19
FEMA Number:
3683
Beilstein:
507689
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.542
NACRES:
NA.21
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Assay

≥98%

refractive index

n20/D 1.427 (lit.)

bp

104 °C/22 mmHg (lit.)

density

0.989 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

berry; green; sweet

SMILES string

CCCC(=O)CC(=O)OCC

InChI

1S/C8H14O3/c1-3-5-7(9)6-8(10)11-4-2/h3-6H2,1-2H3

InChI key

KQWWVLVLVYYYDT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl 3-oxohexanoate is a volatile flavor ester that is reported to occur in banana fruit. It can undergo bioreduction to form ethyl (R)-3-hydroxyhexanoate, another key food flavoring agent.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

194.0 °F - closed cup

Flash Point(C)

90 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Volatile components of banana fruit (Musa sapientum L.)?Indio? from Cuba.
Pino JA, et al.
J. Essent. Oil Res., 15(2), 79-80 (2003)
Highly enantioselective bioreduction of ethyl 3-oxohexanoate.
de Souza Ramos A, et al.
Tetrahedron Letters, 52(46), 6127-6129 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service