Skip to Content
Merck
All Photos(1)

Documents

W341703

Sigma-Aldrich

3-Penten-2-one

≥70%

Synonym(s):

Methyl propenyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=CHCOCH3
CAS Number:
Molecular Weight:
84.12
FEMA Number:
3417
Beilstein:
1633505
EC Number:
Council of Europe no.:
666
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
7.044

biological source

synthetic

grade

Halal
Kosher

vapor pressure

760 mmHg ( 121 °C)

Assay

≥70%

impurities

~30% Mesityl oxide

refractive index

n20/D 1.437 (lit.)

bp

121-124 °C (lit.)

density

0.862 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fishy; fruity

SMILES string

CC(C=CC)=O

InChI

1S/C5H8O/c1-3-4-5(2)6/h3-4H,1-2H3

InChI key

LABTWGUMFABVFG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

69.8 °F - closed cup

Flash Point(C)

21 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service