Skip to Content
Merck
All Photos(1)

Documents

W337501

Sigma-Aldrich

4-Methylthio-2-butanone

≥97%, FG

Synonym(s):

potato butanone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SCH2CH2COCH3
CAS Number:
Molecular Weight:
118.20
FEMA Number:
3375
EC Number:
Council of Europe no.:
11688
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
12.057
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 172.320

Assay

≥97%

refractive index

n20/D 1.473 (lit.)

bp

105-107 °C/55 mmHg (lit.)

density

1.003 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

earthy; vegetable; potato; sulfurous; tomato

SMILES string

CSCCC(C)=O

InChI

1S/C5H10OS/c1-5(6)3-4-7-2/h3-4H2,1-2H3

InChI key

DRGHCRKOWMAZAO-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

161.6 °F - closed cup

Flash Point(C)

72 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service