Skip to Content
Merck
All Photos(1)

Key Documents

W322008

Sigma-Aldrich

2-Phenethylamine

≥99%

Synonym(s):

β-Phenylethylamine, 2-Phenylethylamine

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Linear Formula:
C6H5CH2CH2NH2
CAS Number:
Molecular Weight:
121.18
FEMA Number:
3220
Beilstein:
507488
EC Number:
Council of Europe no.:
708
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
11.006

grade

Kosher

vapor density

4.18 (vs air)

Assay

≥99%

refractive index

n20/D 1.533 (lit.)

bp

197-200 °C (lit.)

density

0.962 g/mL at 20 °C (lit.)

SMILES string

NCCc1ccccc1

InChI

1S/C8H11N/c9-7-6-8-4-2-1-3-5-8/h1-5H,6-7,9H2

InChI key

BHHGXPLMPWCGHP-UHFFFAOYSA-N

Gene Information

human ... AOC3(8639)
mouse ... Aoc3(11754)
rat ... Htr2a(29595)

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

177.8 °F - closed cup

Flash Point(C)

81 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service