Skip to Content
Merck
All Photos(1)

Key Documents

W279801

Sigma-Aldrich

1,1-Dimethoxyoctane

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H3C(CH2)6CH(OCH3)2
CAS Number:
Molecular Weight:
174.28
FEMA Number:
2798
EC Number:
MDL number:
Flavis number:
6.008

biological source

synthetic

Assay

97%

refractive index

n20/D 1.416 (lit.)

bp

206-207 °C (lit.)

density

0.851 g/mL at 25 °C (lit.)

Organoleptic

violet

SMILES string

CCCCCCCC(OC)OC

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1B

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

185.0 °F - closed cup

Flash Point(C)

85 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service