Skip to Content
Merck
All Photos(1)

Key Documents

W272604

Sigma-Aldrich

Methyl 2-nonynoate

99%, FG

Sign Into View Organizational & Contract Pricing

Select a Size

W272604-SAMPLE-K
€50.30
100 G
€120.00
1 KG
€536.00

€50.30


Available to ship on14 April 2025Details


Request a Bulk OrderRequest more information

Select a Size

Change View
W272604-SAMPLE-K
€50.30
100 G
€120.00
1 KG
€536.00

About This Item

Linear Formula:
CH3(CH2)5C≡CCO2CH3
CAS Number:
Molecular Weight:
168.23
FEMA Number:
2726
EC Number:
Council of Europe no.:
479
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.156
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

€50.30


Available to ship on14 April 2025Details


Request a Bulk OrderRequest more information

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002

Assay

99%

refractive index

n20/D 1.448 (lit.)

bp

121 °C/20 mmHg (lit.)

density

0.915 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

green; floral; violet

SMILES string

CCCCCCC#CC(=O)OC

InChI

1S/C10H16O2/c1-3-4-5-6-7-8-9-10(11)12-2/h3-7H2,1-2H3

InChI key

NTLJTUMJJWVCTL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

213.8 °F - closed cup

Flash Point(C)

101 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roman Rieger et al.
Journal of autoimmunity, 27(1), 7-16 (2006-08-01)
Antimitochondrial antibodies (AMA) are unique among autoimmune serologic reactants because of their extremely high association with the index disease primary biliary cirrhosis (PBC). This autoantibody response is specifically directed only to the lipoyl domain of the mitochondrial 2-oxo-acid dehydrogenase complexes
Allergic contact dermatitis from methyl heptine and methyl octine carbonates.
J S English et al.
Contact dermatitis, 18(3), 174-175 (1988-03-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service