W254010
Heptaldehyde
natural, ≥92%
Synonym(s):
Heptanal, 1-Heptanal, Aldehyde C7, Enanthaldehyde, Oenanthaldehyde
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3(CH2)5CHO
CAS Number:
Molecular Weight:
114.19
FEMA Number:
2540
Beilstein:
1560236
EC Number:
Council of Europe no.:
117c
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
5.031
grade
Kosher
natural
reg. compliance
FCC
Assay
≥92%
refractive index
n20/D 1.413 (lit.)
bp
153 °C (lit.)
mp
−43 °C (lit.)
density
0.817 g/mL at 25 °C (lit.)
SMILES string
CCCCCCC=O
InChI
1S/C7H14O/c1-2-3-4-5-6-7-8/h7H,2-6H2,1H3
InChI key
FXHGMKSSBGDXIY-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3 - Flam. Liq. 3 - Skin Irrit. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
95.0 °F - closed cup
Flash Point(C)
35 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Amir Dekel et al.
Scientific reports, 6, 37330-37330 (2016-11-17)
Olfaction is a key insect adaptation to a wide range of habitats. In the last thirty years, the detection of octenol by blood-feeding insects has been primarily understood in the context of animal host-seeking. The recent discovery of a conserved
Nan Li et al.
Toxicology in vitro : an international journal published in association with BIBRA, 41, 159-167 (2017-02-22)
The in vitro EpiSkin™ test method was validated in 2007 by the European Union Reference Laboratory for alternatives to animal testing (EURL ECVAM) as a full replacement method for the Draize acute skin irritation test and adopted in the OECD
C A Mini et al.
Toxicology in vitro : an international journal published in association with BIBRA, 71, 105069-105069 (2020-12-15)
Three-dimensional skin models, also named 3D skin models, human skin equivalents (HSEs), or Human Epidermal Equivalents (HEEs), have been increasingly used for chemical assessments in terms of efficacy and safety. Considering this, we developed an HEE model using immortalized HaCaT
Vincent O Nyasembe et al.
Parasites & vectors, 8, 581-581 (2015-11-11)
Lack of effective vaccines and therapeutics for important arboviral diseases such as Rift Valley fever (RVF) and dengue, necessitates continuous monitoring of vector populations for infections in them. Plant-based lures as surveillance tools has the potential of targeting mosquitoes of
Jie Li et al.
Clinica chimica acta; international journal of clinical chemistry, 436, 59-67 (2014-05-13)
Breast cancer (BC) remains the most commonly diagnosed malignancy in women. We investigated 4 straight aldehydes in the exhaled breath as potential early BC diagnostic biomarkers. End-tailed breath were collected by Bio-VOC® sampler and assayed by gas chromatography-mass spectrometry. Kruskal-Wallis
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service