Skip to Content
Merck
All Photos(1)

Documents

W230405

Sigma-Aldrich

Citral diethyl acetal

≥93%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H26O2
CAS Number:
Molecular Weight:
226.36
FEMA Number:
2304
EC Number:
Council of Europe no.:
38
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
6.004
NACRES:
NA.21

biological source

synthetic

grade

Halal
Kosher

reg. compliance

FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥93%

may contain

<0.10% alpha-tocopherol, synthetic as stabilizer

refractive index

n20/D 1.449 (lit.)

bp

140-142 °C/15 mmHg (lit.)

density

0.875 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

green; lemon; citrus; waxy

SMILES string

CCOC(OCC)\C=C(/C)CC\C=C(/C)C

InChI

1S/C14H26O2/c1-6-15-14(16-7-2)11-13(5)10-8-9-12(3)4/h9,11,14H,6-8,10H2,1-5H3/b13-11+

InChI key

NTXGFKWLJFHGGJ-ACCUITESSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service