Skip to Content
Merck
All Photos(1)

Key Documents

W227102

Sigma-Aldrich

Celery seed oil

natural, FCC, FG

Synonym(s):

Apium graveolens

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2271
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.21

biological source

Apium graveolens L.

grade

FG
Kosher
natural

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 182.20

optical activity

[α]20/D +70.0°, neat

origin

India origin

refractive index

n20/D 1.482 (lit.)

bp

183 °C (lit.)

density

0.88 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

celery

Organoleptic

celery

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

134.6 °F - closed cup

Flash Point(C)

57 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service