THS03118
ThalesNano CatCart® catalyst cartridge system, 30 mm L
1% Pd/Al2O3
Synonym(s):
Heterogeneous catalysis system, safe Heterogeneous catalysis
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
60104500
NACRES:
NB.51
packaging
pkg of 6 ea
manufacturer/tradename
ThalesNano THS 03118
diam.
4 mm
Looking for similar products? Visit Product Comparison Guide
General description
CatCarts are suitable for use only with Thalesnano H-Cube reactors.
Legal Information
CatCart is a registered trademark of ThalesNano Nanotechnology Inc.
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service