Key Documents
SYNF909100H
Synthware pear-shaped Schlenk flask with stopcock
capacity 100 mL, joint: ST/NS 19/22
Synonym(s):
Synthware F909100H, modified Schlenk reaction flask
Select a Size
1 EA
€90.90
€90.90
Check Cart for Availability
Select a Size
Change View
1 EA
€90.90
About This Item
UNSPSC Code:
41121804
NACRES:
NB.51
€90.90
Check Cart for Availability
material
borosilicate glass flask
pear-shaped
single-neck
Joint
joint: ST/NS19/22
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F909100H
capacity
100 mL
General description
A suffix of G indicates a glass stopcock, H indicates a high-vacuum valve, no letter suffix indicates a PTFE Stopcock.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)