SYNF904000G
Synthware™ pear-shaped Schlenk flask with stopcock
capacity 1,000 mL, joint: ST/NS 24/40
Synonym(s):
Synthware F904000G, modified Schlenk reaction flask
All Photos(1)
About This Item
Recommended Products
material
borosilicate glass flask
pear-shaped
single-neck
Joint
joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F904000G
capacity
1,000 mL
Related Categories
General description
A suffix of G indicates a glass stopcock, H indicates a high-vacuum valve, no letter suffix indicates a PTFE Stopcock.
Legal Information
Synthware is a trademark of Kemtech America Inc.