SYNA841420
Synthware™ vacuum filtration adapter
bottom joint: ST/NS 14/20, Stopper Size: #1, stopper size #1
Synonym(s):
Synthware A841420, Glass adapter vacuum filtration
All Photos(1)
About This Item
Recommended Products
Joint
bottom joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
A841420
Synthware A841420
Related Categories
General description
Used with plain stem Buchner funnels and pluro-stopper. Hose connection has an O.D of 10 mm at the largest serration. Order pluro-stoppers below separately.
Legal Information
Synthware is a trademark of Kemtech America Inc.