Skip to Content
Merck
All Photos(1)

Key Documents

SLW718347

Pyrex® Pestle

rough finish, L 125 mm

Synonym(s):

Pyrex, SciLabware 718347

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€22.10

€22.10


Estimated to ship on12 April 2025



Select a Size

Change View
1 EA
€22.10

About This Item

UNSPSC Code:
41121800
NACRES:
NB.35

€22.10


Estimated to ship on12 April 2025


material

borosilicate glass

manufacturer/tradename

718347
SciLabware

L

125 mm

Looking for similar products? Visit Product Comparison Guide

General description

  • Ideal for crushing solids into powders for easier handling
  • Manufactured from Pyrex borosilicate glass for highest resistance to thermal shock and chemical corrosion
  • Available in a rough or glazed finish
  • For use with Pyrex Mortars

Legal Information

PYREX is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service